Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polystyrene-block-poly(ethylene-ran-butylene)-block-polystyrene

Catalog No. 200565250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-056-5250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 20-056-5250G Supplier Sigma Aldrich Supplier No. 200565250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polystyrene-block-poly(ethylene-ran-butylene)-block-polystyrene, also known as SEBS polymer, is used as a thermoplastic elastomer. It has good thermal and chemical stability with tunable mechanical properties.SEBS can be used in the formation of an anion exchange membrane with an energy efficiency of ∼ 86% for the fabrication of redox flow batteries. It can also be used to form sulfonated membranes with high water permeability and thermomechanical stability for HVAC applications. SEBS finds its usage as an ion exchange membrane with a capacity of ∼1.91 mmol g-1 for fuel cells.

Specifications

CAS 66070-58-4
Density 0.91 g/mL (at 25°C)
Linear Formula [-CH2CH(C6H5)-]W(-CH2CH2-)x[-CH2CH(C2H5)-]y[-CH2CH(C6H5)-]Z
Quantity 250 g
MDL Number MFCD00134026
Molecular Weight (g/mol) 89000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.