Learn More
Perkin Elmer US LLC Polystyrene Disposable Sample Cups, 1.5 mL - Pkg. 1000
Catalog No. 502112395
Encompass_Preferred
Sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample.
Catalog No. 50-211-2395 Supplier Perkin Elmer US LLC Supplier No. B0119079
Sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample.