Learn More
Greiner Bio-One Polystyrene Frame for Strips, Non-Sterile
Catalog No. 07000835 Shop All Greiner Bio-One Products
Change view
Click to view available options
Polystyrene Frame for Strips
Catalog No.
07-000-835 Polystyrene Frame for Strips
1 options
Catalog No. 07-000-835 Supplier Greiner Bio-One Supplier No. 754101
Free of detectable DNase/RNase, human DNA and pyrogens.
  • Non-Sterile
  • White
TRUSTED_SUSTAINABILITY
Specifications
Format Strip Plate
Autoclavable Not Autoclavable
Color White
Product Type Frame for strips
Material Polystyrene
Sterility Non-sterile
Dnase Free Yes
No. per Case 400
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C