Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polystyrene for GPC 500, MilliporeSigma™ Supelco™

Catalog No. 111003937 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3937 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3937 Supplier MilliporeSigma Supelco Supplier No. 814011G
Add to Cart
Add to Cart

Polystyrene is a synthetic thermoplastic, which can find a wide range of applications due to its properties like low cost, rigidity, low specific weight, high chemical resistance, mechanical flexibility, biocompatibility and good processability.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Neat
Linear Formula [CH2 CH(C6H5)]n
MDL Number MFCD00084450
Synonym PS
Type Analytical Standard
Quantity 1 g
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.