Alfa Aesar™ Polystyrene latex microsphere, 1 micron, dry form

Manufacturer: Alfa Aesar™ 43711ME

Quantity 500mg

 View more versions of this product

Catalog No. AA43711ME

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9003-53-6
    Molecular Formula [-CH2CH(C6H5)-]n
    MDL Number MFCD00084450
    Chemical Name or Material Polystyrene latex microsphere, Dry form

    Specifications

    Physical Form Microsphere
    Molecular Formula [-CH2CH(C6H5)-]n
    CAS 9003-53-6
    Odor Odorless
    Particle Size 1 Micron
    TSCA TSCA
    MDL Number MFCD00084450
    Quantity 500mg
    Chemical Name or Material Polystyrene latex microsphere, Dry form

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit