Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polystyrene latex microsphere, 2 micron, 10 wt% dispersion in water

Catalog No. AA419187C
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA419187C 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA419187C Supplier Thermo Scientific Chemicals Supplier No. 0419187C
Only null left
Add to Cart

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol

Use in protein binding; size markers. The most popular use of latex beads is in the diagnostic use of adsorbed antibodies or antigens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-53-6
Molecular Formula (C8H8)n
Molecular Weight (g/mol) 104.15
MDL Number MFCD00084450
InChI Key PPBRXRYQALVLMV-UHFFFAOYSA-N
Synonym ethenylbenzene, phenylethylene, vinylbenzene, styrol, benzene, ethenyl, cinnamene, phenylethene, monomer, phenethylene, styrolene
PubChem CID 7501
ChEBI CHEBI:27452
IUPAC Name ethenylbenzene
SMILES *-CC(-*)C1=CC=CC=C1

Specifications

Physical Form Dispersion
Solubility Information Soluble in Benzene,chloroform,carbon tetrachloride,cyclohexane,dimethylformamide,methylene chloride,pyridine,tetrahydrofuran,toluene,xylene. Insoluble in Acetone,butane,ethanol,ether,methanol,hexane,phenol,propanol,water.
Quantity 15 mL
Concentration or Composition (by Analyte or Components) 10 wt% dispersion in water
Odor Odorless
Particle Size 2 Micron
Chemical Name or Material Polystyrene latex microsphere
RTECSNumber WL6475000
TSCA Yes
storageNote1 2°C to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.