Learn More
Corning™ Polystyrene Microplates
GREENER_CHOICE
Catalog No. 07201037 Shop All Corning Products
Change view
Click to view available options
Total Volume (Metric) Well:
190 μL
Surface Treatment:
High Binding
Catalog No. Total Volume (Metric) Well Surface Treatment
07-201-037 190 μL High Binding
1 options
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Catalog No. 07-201-037 Supplier Corning™ Supplier No. 2483
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
Designed for in vitro diagnostic assays
The Stripwell™ low volume microplates allow test kit manufacturers to reduce overall costs, with a reduction in the amount of reagents and antibody necessary to perform assays.
  • Designed with one-half the well area of a 'regular' 8 well strip, providing a much greater surface to volume ratio
  • Recommended working volume of 50μL to 125μL
  • Designed for in vitro diagnostic assays
  • Untreated surface is hydrophobic in nature
  • Color coded strips
  • Bar coding available on request
Maleimide surface covalently couples to sulfhydryl groups (via SH moieties) and are ideal for assays requiring site directed orientation of a particular biomolecule, especially antibodies. Minimal surface preparation is required. Individually packed.
TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Array 8 x 12
Binding Type High Binding
Color Black
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 125 μL
Sterility Non-sterile
Surface Treatment High Binding
Total Volume (Metric) Well 190 μL
No. per Case 100
No. per Pack 25
Well Color Black