Learn More
Spherotech POLYSTYRENE PARTICLE STD KIT
Catalog No. NC9960410
Encompass_Preferred
Polystyrene Particle Size Standard Kit, Flow Cytometry Grade, 0.05% DVB Cross-linked, 2.5 x 1E6/mL, 6x5 mL, 2.0 um, 3.3 um, 5.05 um, 7.66 um, 10.1 um, 15.3 um
Catalog No. NC9960410 Supplier Spherotech Supplier No. PPS6K
Polystyrene Particle Size Standard Kit, Flow Cytometry Grade, 0.05% DVB Cross-linked, 2.5 x 1E6/mL, 6x5 mL, 2.0 um, 3.3 um, 5.05 um, 7.66 um, 10.1 um, 15.3 um