Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer™ Essentials Pipette Basins

Catalog No. 03391539
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
Sterility:
Non-sterile
Sterile
No. per Case:
50
100
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility No. per Case
03-391-539 50 mL Sterile 100
03-391-535 25 mL Sterile 50
03-391-536 25 mL Sterile 100
03-391-537 25 mL Non-sterile 100
03-391-538 50 mL Sterile 50
03-391-540 50 mL Non-sterile 100
03-391-541 100 mL Sterile 50
03-391-542 100 mL Sterile 100
03-391-543 100 mL Non-sterile 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 03-391-539 Supplier Cole-Parmer™ Essentials Supplier No. 0439527
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

V-shaped design minimizes reagent waste

  • Certified RNase-, DNase-, and pyrogen-free

The pipette reagent reservoirs feature a V-shaped bottom, making multichannel pipetting easier and ensuring complete aspiration of costly reagents. They are available in a variety of volumes come in either sterile or nonsterile formats.

Single Chamber Reservoirs are made of white polystyrene and constructed with low cross bars at each end for added stiffness and strength. Each reservoir features graduations on the inner walls, pour spouts in every corner, and one larger spout to facilitate spill-free pouring. Available in 25, 50, and 100 mL volumes, the reservoirs are ideal for 8- and 12-channel pipettors.

The 12-Chamber Reservoir is designed for serial dilutions or pipetting different reagents using a 12-channel pipettor. Each numbered chamber holds up to 3 mL. The chemical-resistant polypropylene reservoir can withstand temperatures from -122 to 250°F (-80 to 121°C). Tight fitting lid minimizes evaporation or contamination during incubation or storage.

Specifications

Color White
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Sterile
Type Pipetting Reservoir
For Use With (Application) Holding Reagent
No. per Case 100
Quantity 100/Cs.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.