Learn More
Bellco Glass, Inc. Polystyrene Plasteur Pipet, 9 Inch Length, Bulk Packed, Sterile
SDP
Catalog No. 502329840
Encompass_Preferred
DescriptionThe Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plasticFine tip is ideal for precision aspiratingNo more broken glass: more durable than standard glass pipetsNo damage during shipping and handling as compared to traditional glass pipetsAvailable sterile for convenience Pack Description25 Bulk Packed/BagCase Quantity500SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneGraduatedNoRNase/DNase FreeYes
Catalog No. 50-232-9840 Supplier Bellco Glass, Inc. Supplier No. 229285000
Add to Cart
Add to Cart
DescriptionThe Plasteur Plastic Pasteur Pipet offers the aspirating precision of a glass pipet and the safety benefits of plasticFine tip is ideal for precision aspiratingNo more broken glass: more durable than standard glass pipetsNo damage during shipping and handling as compared to traditional glass pipetsAvailable sterile for convenience Pack Description25 Bulk Packed/BagCase Quantity500SterileYesNon-pyrogenicYesPrimary ColorClearPrimary MaterialPolystyreneGraduatedNoRNase/DNase FreeYes