Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polystyrene Sign: NOTICE - MATERIAL SAFETY DATA SHEETS & THE WRITTEN HAZARD COMMUNICATION PROGRAM LOCATED HERE

Catalog No. p-7122403 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
10 x 7 in.
14 x 10 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19-105-940 14 x 10 in.
19-105-938 10 x 7 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-105-940 Supplier Brady™ Supplier No. 126472

Improve workplace safety with proper signage and markings. This Brady™ polystyrene sign is a durable, clear and effective tool to raise awareness to workplace hazards.

  • Polystyrene material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Polystyrene signs are thin, pliable and durable
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use

Specifications

Material Polystyrene
Shape Rectangular
Dimensions (L x W) 14 x 10 in.
Displayed Text NOTICE - MATERIAL SAFETY DATA SHEETS & THE WRITTEN HAZARD COMMUNICATION PROGRAM LOCATED HERE
Color Blue on White

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.