Learn More
Waters Corp Waters Polystyrene Standard, 10,100 Da, GPC, 5 g
Catalog No. 50785024 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
An individual GPC (Gel Permeation Chromatography) standard for molecular weight determination. It consists of Polystyrene at 10,100 Da and is used for characterizing or confirming chemical analysis. This high-quality, proven standard provides confidence in results. The Polystyrene Standard 10,100 is provided in a single package and can be stored in a refrigerator at 4°C. Its shelf life is 36 months, but only one week after reconstitution.
  • Individual GPC standard for molecular weight determination.
  • Polystyrene at 10,100 Da.
  • High-quality, proven standard.
  • Designed to provide known molecular weight information.
  • Traceability of performance-based reference materials.
Catalog No. 50-785-024 Supplier Waters Corp Supplier No. WAT011600
An individual GPC (Gel Permeation Chromatography) standard for molecular weight determination. It consists of Polystyrene at 10,100 Da and is used for characterizing or confirming chemical analysis. This high-quality, proven standard provides confidence in results. The Polystyrene Standard 10,100 is provided in a single package and can be stored in a refrigerator at 4°C. Its shelf life is 36 months, but only one week after reconstitution.
  • Individual GPC standard for molecular weight determination.
  • Polystyrene at 10,100 Da.
  • High-quality, proven standard.
  • Designed to provide known molecular weight information.
  • Traceability of performance-based reference materials.