Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Polystyrene Standard, 110,000 Da, Gel Permeation Chromatography, Calibration

Catalog No. 50785027 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This individual GPC standard is used for molecular weight determination, consisting of Polystyrene at 110,000 Da. These high-quality, proven polystyrene calibration standards are designed for characterizing or confirming chemical analysis, providing confidence in results.

  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: Room temperature
  • System type: LC
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Mass: 5 g
  • Type: Calibration
  • Application: Gel permeation
  • Units per package: 1 pk

Catalog No. 50-785-027 Supplier Waters Corp Supplier No. WAT011606
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This individual GPC standard is used for molecular weight determination, consisting of Polystyrene at 110,000 Da. These high-quality, proven polystyrene calibration standards are designed for characterizing or confirming chemical analysis, providing confidence in results.

  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: Room temperature
  • System type: LC
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Mass: 5 g
  • Type: Calibration
  • Application: Gel permeation
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.