Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polystyrene Standard, 1,300,000 Da, GPC, Calibration

Catalog No. 50785032 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This individual GPC standard is designed for molecular weight determination. It is a Polystyrene calibration standard with a mass of 1,300,000 Da, used for characterizing or confirming chemical analysis, providing confidence in results.

  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: Room temperature
  • System type: LC
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Mass: 1 g
  • Type: Calibration
  • Application: Gel permeation
  • Units per package: 1 pk

Catalog No. 50-785-032 Supplier Waters Corp Supplier No. WAT011616
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This individual GPC standard is designed for molecular weight determination. It is a Polystyrene calibration standard with a mass of 1,300,000 Da, used for characterizing or confirming chemical analysis, providing confidence in results.

  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: Room temperature
  • System type: LC
  • Shelf life: 36 months
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Mass: 1 g
  • Type: Calibration
  • Application: Gel permeation
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.