Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polystyrene Standard, 17,000 Da, 5 g

Catalog No. 50785025 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

An individual GPC standard (Gel Permeation Chromatography standards) used for molecular weight determination. These polystyrene calibration standards are for use in characterizing or confirming chemical analysis, providing high-quality, proven standards to ensure confidence in results.

  • Molecular weight of 17,000 Da
  • Mass of 5 g
  • One unit per package
  • Tolerance of 10% (Mp)
  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Solid formulation
  • Store at room temperature
  • Compatible with LC system types
  • Shelf life of 36 months
  • Ambient shipping condition
  • Type: Calibration standard
  • Application: Gel permeation

Catalog No. 50-785-025 Supplier Waters Corp Supplier No. WAT011602
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An individual GPC standard (Gel Permeation Chromatography standards) used for molecular weight determination. These polystyrene calibration standards are for use in characterizing or confirming chemical analysis, providing high-quality, proven standards to ensure confidence in results.

  • Molecular weight of 17,000 Da
  • Mass of 5 g
  • One unit per package
  • Tolerance of 10% (Mp)
  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Solid formulation
  • Store at room temperature
  • Compatible with LC system types
  • Shelf life of 36 months
  • Ambient shipping condition
  • Type: Calibration standard
  • Application: Gel permeation

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.