Learn More
Waters Corp Waters Corporation Polystyrene Standard, 180,000 Da, GPC, Calibration Standard
Catalog No. 50785028 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is an individual Gel Permeation Chromatography (GPC) standard used for molecular weight determination. It consists of Polystyrene at 180,000 Da and serves as a high-quality calibration standard for characterizing or confirming chemical analyses.
  • Molecular weight of 180,000 Da.
  • Suitable for gel permeation chromatography (GPC/GFC/SEC) applications.
  • Compatible with UV, RI/ELSD detection methods.
  • Provided in solid formulation.
  • Can be stored at room temperature.
  • Has a shelf life of 36 months.
  • Ships under ambient conditions.
  • Provides a tolerance of 10% (Mp).
  • Supplied as 5 g mass.
  • Functions as a calibration standard.
  • One unit per package.
Catalog No. 50-785-028 Supplier Waters Corp Supplier No. WAT011608
This product is an individual Gel Permeation Chromatography (GPC) standard used for molecular weight determination. It consists of Polystyrene at 180,000 Da and serves as a high-quality calibration standard for characterizing or confirming chemical analyses.
  • Molecular weight of 180,000 Da.
  • Suitable for gel permeation chromatography (GPC/GFC/SEC) applications.
  • Compatible with UV, RI/ELSD detection methods.
  • Provided in solid formulation.
  • Can be stored at room temperature.
  • Has a shelf life of 36 months.
  • Ships under ambient conditions.
  • Provides a tolerance of 10% (Mp).
  • Supplied as 5 g mass.
  • Functions as a calibration standard.
  • One unit per package.