Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polystyrene Standard, 2,800,000 Da, Gel Permeation Chromatography, Calibration

Catalog No. 50785033 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Polystyrene Standard, with a molecular weight of 2,800,000 Da, serves as an individual GPC (Gel Permeation Chromatography) standard for precise molecular weight determination. It's designed to characterize and confirm chemical analyses, ensuring confidence in your results.

  • Individual GPC standard for molecular weight determination.
  • Polystyrene with a molecular weight of 2,800,000 Da.
  • Calibration standards for chemical analysis.
  • Separation mode: GPC/GFC/SEC.
  • Detection method: UV, RI/ELSD.
  • Formulation: Solid.
  • Storage condition: 4°C (Fridge).
  • Shelf life: 36 months.
  • Mass: 1 g.

Catalog No. 50-785-033 Supplier Waters Corp Supplier No. WAT011618
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Polystyrene Standard, with a molecular weight of 2,800,000 Da, serves as an individual GPC (Gel Permeation Chromatography) standard for precise molecular weight determination. It's designed to characterize and confirm chemical analyses, ensuring confidence in your results.

  • Individual GPC standard for molecular weight determination.
  • Polystyrene with a molecular weight of 2,800,000 Da.
  • Calibration standards for chemical analysis.
  • Separation mode: GPC/GFC/SEC.
  • Detection method: UV, RI/ELSD.
  • Formulation: Solid.
  • Storage condition: 4°C (Fridge).
  • Shelf life: 36 months.
  • Mass: 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.