Learn More
Waters Corp Waters Corporation Polystyrene Standard, 3,600,000 MW, 1 g/vial
Catalog No. 50785034 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a polystyrene standard designed for use in gel permeation chromatography (GPC) applications. It provides a reference for molecular weight determination, essential for accurate analysis of polymers.
  • Polystyrene standard with an approximate molecular weight of 3,600,000.
  • Supplied as 1 g per vial for convenient use.
  • Ideal for calibration in GPC systems.
Catalog No. 50-785-034 Supplier Waters Corp Supplier No. WAT011620
This product is a polystyrene standard designed for use in gel permeation chromatography (GPC) applications. It provides a reference for molecular weight determination, essential for accurate analysis of polymers.
  • Polystyrene standard with an approximate molecular weight of 3,600,000.
  • Supplied as 1 g per vial for convenient use.
  • Ideal for calibration in GPC systems.