Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polystyrene Standard, 4,300,000 Da, GPC/GFC/SEC, Solid

Catalog No. 50785035 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Polystyrene Standard 4,300,000 is an individual GPC standard for accurate molecular weight determination. With a molecular weight of 4,300,000 Da, this high-quality standard helps characterize and confirm chemical analyses, providing confidence in your results.

  • Compatible with GPC, GFC, and SEC applications.
  • Suitable for UV, RI, and ELSD detection methods.
  • Supplied in solid form.
  • Store at 4°C (refrigerated).
  • Designed for Liquid Chromatography (LC) systems.
  • Shelf life of 36 months.
  • Stable for 1 week after reconstitution.
  • Shipped at ambient temperature.
  • Tolerance: ±10% (Mp).
  • Mass: 1 g.
  • Calibration standard type.
  • Used for Gel Permeation applications for precise calibration and reliable molecular weight determination.
  • 1 unit per package.

Catalog No. 50-785-035 Supplier Waters Corp Supplier No. WAT011622
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Polystyrene Standard 4,300,000 is an individual GPC standard for accurate molecular weight determination. With a molecular weight of 4,300,000 Da, this high-quality standard helps characterize and confirm chemical analyses, providing confidence in your results.

  • Compatible with GPC, GFC, and SEC applications.
  • Suitable for UV, RI, and ELSD detection methods.
  • Supplied in solid form.
  • Store at 4°C (refrigerated).
  • Designed for Liquid Chromatography (LC) systems.
  • Shelf life of 36 months.
  • Stable for 1 week after reconstitution.
  • Shipped at ambient temperature.
  • Tolerance: ±10% (Mp).
  • Mass: 1 g.
  • Calibration standard type.
  • Used for Gel Permeation applications for precise calibration and reliable molecular weight determination.
  • 1 unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.