Learn More
Waters Corp WATERS Polystyrene Standard, 4000 Da, Gel Permeation Chromatography
Catalog No. 50785019 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Polystyrene Standard is an individual GPC standard used for molecular weight determination. It has a molecular weight of 4000 Da and is designed for calibrating and confirming chemical analysis, providing confidence in results.
  • High-quality, proven standards ensure confident results.
  • Supports GPC, GFC, and SEC separation modes.
  • Compatible with UV, RI/ELSD detection methods.
  • Solid formulation.
  • Store at 4°C.
  • Designed for LC system types.
  • 36 months shelf life.
  • 1 week shelf life after reconstitution.
  • Ambient shipping condition.
  • 10% (Mp) tolerance.
  • 10 g mass.
  • Calibration type.
  • For gel permeation applications.
  • 1 unit per package.
Catalog No. 50-785-019 Supplier Waters Corp Supplier No. WAT011590
This Polystyrene Standard is an individual GPC standard used for molecular weight determination. It has a molecular weight of 4000 Da and is designed for calibrating and confirming chemical analysis, providing confidence in results.
  • High-quality, proven standards ensure confident results.
  • Supports GPC, GFC, and SEC separation modes.
  • Compatible with UV, RI/ELSD detection methods.
  • Solid formulation.
  • Store at 4°C.
  • Designed for LC system types.
  • 36 months shelf life.
  • 1 week shelf life after reconstitution.
  • Ambient shipping condition.
  • 10% (Mp) tolerance.
  • 10 g mass.
  • Calibration type.
  • For gel permeation applications.
  • 1 unit per package.