Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Polystyrene Standard, 530 Da, For GPC

Catalog No. 50785020 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This individual GPC standard is used for molecular weight determination. It is a polystyrene standard at 530 Da, suitable for calibrating and confirming chemical analyses.

  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: 4°C (fridge)
  • System type: LC
  • Shelf life: 36 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Mass: 10 g
  • Type: Calibration
  • Application: Gel permeation
  • Product type: Standards
  • Units per package: 1 pk

Catalog No. 50-785-020 Supplier Waters Corp Supplier No. WAT011592
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This individual GPC standard is used for molecular weight determination. It is a polystyrene standard at 530 Da, suitable for calibrating and confirming chemical analyses.

  • Separation mode: GPC/GFC/SEC
  • Detection method: UV, RI/ELSD
  • Formulation: Solid
  • Storage condition: 4°C (fridge)
  • System type: LC
  • Shelf life: 36 months
  • Shelf life upon reconstitution: 1 week
  • Shipping condition: Ambient
  • Tolerance: 10% (Mp)
  • Mass: 10 g
  • Type: Calibration
  • Application: Gel permeation
  • Product type: Standards
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.