Learn More
Waters Corp Polystyrene Standard 6,400
Catalog No. 50785023 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
An individual GPC standard (Gel Permeation Chromatography standards) for molecular weight determination. Polystyrene at 6,400 Da. Polystyrene calibration standards for use in characterizing or confirming your chemical analysis. This high-quality, proven standards provide confidence in your results.
Catalog No. 50-785-023 Supplier Waters Corp Supplier No. WAT011598
An individual GPC standard (Gel Permeation Chromatography standards) for molecular weight determination. Polystyrene at 6,400 Da. Polystyrene calibration standards for use in characterizing or confirming your chemical analysis. This high-quality, proven standards provide confidence in your results.