Learn More
Waters Corp Waters Corporation Polystyrene Standard, 780,000 Da, 5 g, Calibration
Catalog No. 50785031 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This individual Gel Permeation Chromatography (GPC) standard, consisting of Polystyrene with a molecular weight of 780,000 Da, is designed for molecular weight determination. These calibration standards are used for characterizing or confirming chemical analysis.
  • Individual GPC standard for molecular weight determination
  • Consists of Polystyrene with a molecular weight of 780,000 Da
  • Used for characterizing or confirming chemical analysis
  • Provides high-quality, proven standards for confidence in results
  • Supports GPC/GFC/SEC separation modes
  • Compatible with UV, RI/ELSD detection methods
  • Solid formulation
  • Shelf life of 36 months
  • 5 g mass
Catalog No. 50-785-031 Supplier Waters Corp Supplier No. WAT011614
This individual Gel Permeation Chromatography (GPC) standard, consisting of Polystyrene with a molecular weight of 780,000 Da, is designed for molecular weight determination. These calibration standards are used for characterizing or confirming chemical analysis.
  • Individual GPC standard for molecular weight determination
  • Consists of Polystyrene with a molecular weight of 780,000 Da
  • Used for characterizing or confirming chemical analysis
  • Provides high-quality, proven standards for confidence in results
  • Supports GPC/GFC/SEC separation modes
  • Compatible with UV, RI/ELSD detection methods
  • Solid formulation
  • Shelf life of 36 months
  • 5 g mass