Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Polystyrene Standard, 8,400,000 Da, GPC, Calibration

Catalog No. 50785038 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Polystyrene Standard is an individual Gel Permeation Chromatography (GPC) calibration standard designed for precise molecular weight determination. With a molecular weight of 8,400,000 Daltons, it's suitable for characterization and separation of large polymeric compounds and biomolecules. This solid-form standard ensures excellent reproducibility across various detection methods like Refractive Index (RI), Evaporative Light Scattering Detection (ELSD), and UV, for LC-based GPC/SEC workflows.

  • Molecular weight of 8,400,000 Daltons.
  • Supports GPC/GFC/SEC separation modes.
  • UV, RI, and ELSD detection compatible.
  • Solid formulation.
  • Storage condition: 4°C.
  • Shelf life: 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient.
  • Mass: 1 g.
  • Units per package: 1 pk.

Catalog No. 50-785-038 Supplier Waters Corp Supplier No. WAT011628
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Polystyrene Standard is an individual Gel Permeation Chromatography (GPC) calibration standard designed for precise molecular weight determination. With a molecular weight of 8,400,000 Daltons, it's suitable for characterization and separation of large polymeric compounds and biomolecules. This solid-form standard ensures excellent reproducibility across various detection methods like Refractive Index (RI), Evaporative Light Scattering Detection (ELSD), and UV, for LC-based GPC/SEC workflows.

  • Molecular weight of 8,400,000 Daltons.
  • Supports GPC/GFC/SEC separation modes.
  • UV, RI, and ELSD detection compatible.
  • Solid formulation.
  • Storage condition: 4°C.
  • Shelf life: 36 months.
  • Shelf life upon reconstitution: 1 week.
  • Shipping condition: Ambient.
  • Mass: 1 g.
  • Units per package: 1 pk.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.