Learn More
Waters Corp Waters Polystyrene Standards Kit, Mid-High MW, ReadyCal
Catalog No. 50785029 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Polystyrene Standards Kit, Mid-High MW, is designed for calibrating GPC/SEC systems. These ReadyCal standards are pre-weighed and come with a certificate of analysis, providing essential molecular weight data for accurate system calibration.
  • Pre-weighed for ease of use.
  • Includes a certificate of analysis providing MW, Mn, and polydispersity values.
  • Available with 5, 10, or 12 standards in each mixture.
  • Packaged in various configurations, including 5 vials with 5 standards each, 1 vial with 5 standards, or a 10-point kit with 10 vials.
  • Suitable for generating calibration curves for GPC/SEC systems.
  • Molecular weight range from 1000 to 1000000 Da.
Catalog No. 50-785-029 Supplier Waters Corp Supplier No. WAT011610
The Polystyrene Standards Kit, Mid-High MW, is designed for calibrating GPC/SEC systems. These ReadyCal standards are pre-weighed and come with a certificate of analysis, providing essential molecular weight data for accurate system calibration.
  • Pre-weighed for ease of use.
  • Includes a certificate of analysis providing MW, Mn, and polydispersity values.
  • Available with 5, 10, or 12 standards in each mixture.
  • Packaged in various configurations, including 5 vials with 5 standards each, 1 vial with 5 standards, or a 10-point kit with 10 vials.
  • Suitable for generating calibration curves for GPC/SEC systems.
  • Molecular weight range from 1000 to 1000000 Da.