Learn More
Waters Corp Waters GPC Standards Kit, Polystyrene, 10 Standards, 0.5 g Each
Catalog No. 50785341 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Polystyrene Mid MW Standards Kit, also known as the Polystyrene Kit SM-105, is an Organic Gel Permeation Chromatography (GPC) Standards Kit used for the calibration of organic GPC columns. This kit is utilized for non-aqueous molecular weight determination and contains polymer-specific calibration standards tailored for polystyrene analysis. Each standard is shipped with 500 mg in a vial, providing a quick and reliable reference with known molecular weight ranges.
  • Used for the calibration of organic GPC columns.
  • Suitable for non-aqueous molecular weight determination.
  • Contains polymer-specific calibration standards for polystyrene analysis.
  • Includes 10 different molecular weight standards (1200 to 2,750,000).
  • Each standard contains 500 mg (0.5 g) of material.
  • Provides a quick and reliable reference.
Catalog No. 50-785-341 Supplier Waters Corp Supplier No. WAT034209
The Polystyrene Mid MW Standards Kit, also known as the Polystyrene Kit SM-105, is an Organic Gel Permeation Chromatography (GPC) Standards Kit used for the calibration of organic GPC columns. This kit is utilized for non-aqueous molecular weight determination and contains polymer-specific calibration standards tailored for polystyrene analysis. Each standard is shipped with 500 mg in a vial, providing a quick and reliable reference with known molecular weight ranges.
  • Used for the calibration of organic GPC columns.
  • Suitable for non-aqueous molecular weight determination.
  • Contains polymer-specific calibration standards for polystyrene analysis.
  • Includes 10 different molecular weight standards (1200 to 2,750,000).
  • Each standard contains 500 mg (0.5 g) of material.
  • Provides a quick and reliable reference.