Learn More
Medchemexpress LLC Polystyrene sulfonic acid | 28210-41-5 | 100 G
SDP
Catalog No. 5000363693
Encompass_Preferred
Polystyrene sulfonic acid (30% in water) is a potentially toxic hazardous substance. It is for research use only and not sold to patients. MedChemExpress offers these Polystyrene products in solution packaging.
  • Harmful to zebrafish hearts and induces male reproductive toxicity in mice
  • PS-MPs at 0.5 μm, 4 μm, and 10 μm particle sizes can enter three testicular cell lines (GC-1 cell line) in vitro
  • PS-MPs lead to decreased sperm quality and testosterone levels in mice
  • PS-MPs accumulate in the testis of mice
  • Induces testicular inflammation and disruption of the blood-testis barrier in mice
Catalog No. 50-003-63693 Supplier Medchemexpress LLC Supplier No. HY148163100G
Polystyrene sulfonic acid (30% in water) is a potentially toxic hazardous substance. It is for research use only and not sold to patients. MedChemExpress offers these Polystyrene products in solution packaging.
  • Harmful to zebrafish hearts and induces male reproductive toxicity in mice
  • PS-MPs at 0.5 μm, 4 μm, and 10 μm particle sizes can enter three testicular cell lines (GC-1 cell line) in vitro
  • PS-MPs lead to decreased sperm quality and testosterone levels in mice
  • PS-MPs accumulate in the testis of mice
  • Induces testicular inflammation and disruption of the blood-testis barrier in mice