Learn More
Medchemexpress LLC Polystyrene sulfonic acid | 28210-41-5 | 30% | 50 G
SDP
Supplier:  Medchemexpress LLC HY14816350G
Encompass_Preferred
Polystyrene sulfonic acid (30% in water) is provided as a potentially toxic hazardous substance, available in solution packaging for research use only. In vitro and in vivo studies have shown that polystyrene-derived microplastics (PS-MPs) can enter testicular cells, leading to decreased sperm quality, testosterone levels, and testicular inflammation in mice. Furthermore, PS-MPs have been observed to cause adverse effects on zebrafish activity, oxidative stress, metabolic changes, and contraction parameters in heart tissue.
  • Suitable for research applications only
  • Provided in solution packaging
  • Studies indicate potential for male reproductive toxicity in mice, including decreased sperm quality and testosterone levels
  • Demonstrates adverse effects on zebrafish activity and heart tissue, including oxidative stress and metabolic changes
  • PS-MPs with specific particle sizes have been shown to accumulate in the testis of mice
Catalog No. 50-003-60582