Learn More
Medchemexpress LLC Polystyrene sulfonic acid | 28210-41-5 | 500 G
SDP
Catalog No. 5000363162
Encompass_Preferred
Polystyrene sulfonic acid (30% in water) is a chemical offered in solution packaging. Research indicates that polystyrene-derived microplastics (PS-MPs) may be harmful, with studies showing adverse effects on zebrafish hearts and induction of male reproductive toxicity in mice.
  • Microplastics derived from polystyrene sulfonic acid can enter testicular cell lines in vitro.
  • May lead to decreased sperm quality and testosterone levels in mice.
  • Demonstrated adverse effects on zebrafish activity and heart tissue contraction.
  • Observed to accumulate in the testis of mice.
  • Induced testicular inflammation and disruption of the blood-testis barrier in mice.
Catalog No. 50-003-63162 Supplier Medchemexpress LLC Supplier No. HY148163500G
Polystyrene sulfonic acid (30% in water) is a chemical offered in solution packaging. Research indicates that polystyrene-derived microplastics (PS-MPs) may be harmful, with studies showing adverse effects on zebrafish hearts and induction of male reproductive toxicity in mice.
  • Microplastics derived from polystyrene sulfonic acid can enter testicular cell lines in vitro.
  • May lead to decreased sperm quality and testosterone levels in mice.
  • Demonstrated adverse effects on zebrafish activity and heart tissue contraction.
  • Observed to accumulate in the testis of mice.
  • Induced testicular inflammation and disruption of the blood-testis barrier in mice.