Learn More
Abcam Polysucrose 400, synthetic sucrose polymer, 25G
Catalog No. 502704570
Encompass_Preferred
MW 0 g/mol. Synthetic high molecular weight polymer made by the copolymerization of sucrose and epichlorohydrin. The molecules have a branched structure with a high content of hydroxyl groups giving a good solubility in aqueous solutions. Used for the density gradient separation of cells, cell membranes and virus cells. It can also be used to isolate different organelles. Has been used in a study to develop a new method for the purification of human motile spermatozoa.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4570 Supplier Abcam Supplier No. AB14657025G
MW 0 g/mol. Synthetic high molecular weight polymer made by the copolymerization of sucrose and epichlorohydrin. The molecules have a branched structure with a high content of hydroxyl groups giving a good solubility in aqueous solutions. Used for the density gradient separation of cells, cell membranes and virus cells. It can also be used to isolate different organelles. Has been used in a study to develop a new method for the purification of human motile spermatozoa.
The product is subject to the following: Abcam Restricted Use Statement