Learn More
Abcam Polysucrose 70, synthetic sucrose polymer, 25G
Catalog No. 502704569
Encompass_Preferred
MW 70000 g/mol. Sucrose-polymer formed by copolymerization of sucrose with epichlorohydrin. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4569 Supplier Abcam Supplier No. AB14657125G
MW 70000 g/mol. Sucrose-polymer formed by copolymerization of sucrose with epichlorohydrin. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement