Learn More
COLDER PRODUCTS COMPANY CPC Back-to-Back Body Adapter Amber tint Polysulfone USP Class VI ADCF Pressure Up to 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F Temperature Max 149 deg C
Catalog No. 503131466
Encompass_Preferred
CPC Back-to-Back Body Adapter Amber tint Polysulfone USP Class VI ADCF Pressure Up to 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F Temperature Max 149 deg C
Catalog No. 50-313-1466 Supplier COLDER PRODUCTS COMPANY Supplier No. MPC17X1739
CPC Back-to-Back Body Adapter Amber tint Polysulfone USP Class VI ADCF Pressure Up to 60 psi 4 1 bar Sterilization Method Gamma Autoclave Temperature Max 300 deg F Temperature Min -40 deg F Temperature Max 149 deg C