Learn More
Advantec MFS, Inc Polysulfone Filter Holders 47mm
Supplier:  Advantec MFS, Inc 43301030
Encompass_Preferred
Polysulfone Filter Holders are durable and economical. Recessed filter support protects membrane from tearing. Simple twist motion connects funnel to base. 300mL Funnel; 8b stopper adapts to flask or manifold; Autoclavable. (other options)
Catalog No. NC0520967