Learn More
Tisch Scientific Polytetrafluoroethylene PTFE Hydrophilic Syringe Filters, 0.22um, 25mm, Luer-Lok/Luer Slip, Sterile, 100 per pack
SDP
Catalog No. 502251501
Encompass
Polytetrafluoroethylene PTFE Hydrophilic Syringe Filters, 0.22um, 25mm, Luer-Lok/Luer Slip, Sterile, 100 per pack
This item is not returnable. View return policy
Catalog No. 50-225-1501 Supplier Tisch Scientific Supplier No. SF18238
Polytetrafluoroethylene PTFE Hydrophilic Syringe Filters, 0.22um, 25mm, Luer-Lok/Luer Slip, Sterile, 100 per pack