Learn More
Polyscience polytherm PAG140
SDP
Catalog No. NC1876720
Encompass_Preferred
PolyScience™ polytherm PAG140PolyScience polytherm PAG140 offers excellent properties for mid-range temperature baths and circulation systems. Polyalkylene glycol is a low-cost alternative to silicone fluids. Ideal for use in stainless steel and polycarbonate tanks.Temperature Range: 50° to 140°C (122° to 284°F)Quantity: 1 gallon (3.8L)Features:• Mid-range oil• Low-cost alternative to silicone fluids• Ideal for use in stainless steel and polycarbonate tanks.
Catalog No. NC1876720 Supplier Polyscience Supplier No. 060350
PolyScience™ polytherm PAG140PolyScience polytherm PAG140 offers excellent properties for mid-range temperature baths and circulation systems. Polyalkylene glycol is a low-cost alternative to silicone fluids. Ideal for use in stainless steel and polycarbonate tanks.Temperature Range: 50° to 140°C (122° to 284°F)Quantity: 1 gallon (3.8L)Features:• Mid-range oil• Low-cost alternative to silicone fluids• Ideal for use in stainless steel and polycarbonate tanks.