Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Polythylene Dropper Bottle
SCI_ED

Catalog No. S37063 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
29.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S37063 29.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37063
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Economy dropping bottle for a controlled dispensing of liquids from LDPE squeeze bottle.

Specifications

Capacity (Metric) 29.5 mL
Capacity (English) 1 oz.
Product Type Dropping Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.