Learn More
Enzo Life Sciences Polyubiquitin (K63-linkage-specific) monoclonal antibody (HWA4C4) (HRP conjugate) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLPW06050100
Encompass_Preferred
Clone: HWA4C4. Isotype: IgG2a. Immunogen: Synthetic peptide containing the ubiquitin K63-isopeptide motif. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Species independent. Specificity: Recognizes K63-linked polyubiquitin in wide range of species. Applications: WB. Purity: Protein G-affinity purified. Formulation: Liquid. In 1X PBS, pH 7.2.
Catalog No. 50-201-1549