Learn More
Enzo Life Sciences Polyubiquitin chains (Ub2-7), (linear) (recombinant) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLUW10100100
Encompass_Preferred
Linear Polyubiquitin chains Ub2-Ub7 Linear Polyubiquitin chains Ub2-Ub7 Purity: ≥95% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS-HCl, pH 8.0, containing 50mM sodium chloride. Source: Produced in E. coli. Long Term Storage: -20°C.
Catalog No. 50-201-2277