Learn More
Cayman Chemical Polyunsaturated Fatty Acid MaxSpec LC-MS Mixture, Purity: ≥98%, 1 ea
SDP
Catalog No. 502967787
Encompass_Preferred
Contains various polyunsaturated fatty acids, including many important essential fatty acids; designed for direct use in LC-MS applications. Each ampule contains at least 1 ml:Adrenic Acid (Item No. 90300) Arachidonic Acid (Item No. 90010) Dihomo-γ-Linolenic Acid (Item No. 90230) Docosahexaenoic Acid (Item No. 90310) Docosapentaenoic Acid (Item No. 90165) Eicosapentaenoic Acid (Item No. 90110) Linoleic Acid (Item No. 90150) α-Linoleic Acid (Item No. 90210) γ-Linoleic Acid (Item No. 90220) Stearidonic Acid (Item No. 90320) (10 µg/ml each supplied in ethanol)
Catalog No. 50-296-7787 Supplier Cayman Chemical Supplier No. 179411 EA
Contains various polyunsaturated fatty acids, including many important essential fatty acids; designed for direct use in LC-MS applications. Each ampule contains at least 1 ml:Adrenic Acid (Item No. 90300) Arachidonic Acid (Item No. 90010) Dihomo-γ-Linolenic Acid (Item No. 90230) Docosahexaenoic Acid (Item No. 90310) Docosapentaenoic Acid (Item No. 90165) Eicosapentaenoic Acid (Item No. 90110) Linoleic Acid (Item No. 90150) α-Linoleic Acid (Item No. 90210) γ-Linoleic Acid (Item No. 90220) Stearidonic Acid (Item No. 90320) (10 µg/ml each supplied in ethanol)