Learn More
PSI URETHANES INC POLYURETHANE BARREL PATCH 11'
Catalog No. NC2137645
Encompass
POLYURETHANE BARREL PATCHESPart Number: 11ft BP-2,PSI is the original and only manufacturer of urethane barrel patch spill control products designed to stop leaks fast on 55-gallon drums. The barrel patches are made of tough, chemical resistant urethane that forms a tight seal.
Catalog No. NC2137645 Supplier PSI URETHANES INC Supplier No. BP2 11FT
POLYURETHANE BARREL PATCHESPart Number: 11ft BP-2,PSI is the original and only manufacturer of urethane barrel patch spill control products designed to stop leaks fast on 55-gallon drums. The barrel patches are made of tough, chemical resistant urethane that forms a tight seal.