Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Polyurethane Foam Chemical Kit
GREENER_CHOICESCI_ED

Catalog No. S73009 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Class
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S73009 1 Class
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S73009 Supplier Innovating Science™ Supplier No. IS7018
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Two premixed chemicals and instructions

Requires

Plastic or paper cups and safety equipment

Excellent demonstration of thermoplastic polymers.

  • Produce polyurethane foam with the condensation reaction of a polyisocyanate and a hydroxyl-containing material (i.e., a polyol derived from propylene oxide).
  • Mix two viscous liquids to initiate a reaction.
  • Lightweight foam expands to 30 times the original volume of the liquids.
  • After about 10 minutes, the foam hardens.
  • Perform in a fume hood and wear chemical splash goggles, apron and gloves as the chemical components are eye and skin irritants.

Specifications

Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Excellent demonstration of thermoplastic polymers
Includes 1 x 500 mL polyurethane foam system, part a (mixture), 1 x 500 mL polyurethane foam system, part b (mixture)
Quantity 1
Dimensions (L x W x H) 9.1875 x 2.625 x 8.25 in.
Class Size 1 Class
Product Type Polyurethane Foam Chemical Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.