Learn More
ACL Inc Staticide 2.75" Polyurethane Foam swabs with round tip. 500 swabs per bag
Catalog No. NC1841776
Encompass_Preferred
Medical grade foam. No adhesives or binders. Non-abradive. High solvent capacity.Little to no particles. Abrasion resistant. ESD Rigid Handle. Small Round Tip. ISO Class 4-7 compatible. Class 10 cleanroom laundered.
Catalog No. NC1841776 Supplier ACL Inc Supplier No. 7020
Medical grade foam. No adhesives or binders. Non-abradive. High solvent capacity.Little to no particles. Abrasion resistant. ESD Rigid Handle. Small Round Tip. ISO Class 4-7 compatible. Class 10 cleanroom laundered.