Learn More
ACL Inc Staticide 5" Polyurethane Foam swabs with wide paddle tip. 100 swabs per bag
Catalog No. NC1841778
Encompass_Preferred
Medical grade foam. No adhesives or binders. Non-abradive. High solvent capacity.Little to no particles. Abrasion resistant. ESD Rigid Handle. Wide Paddle Tip. ISO Class 4-7 compatible. Class 10 cleanroom laundered.
Catalog No. NC1841778 Supplier ACL Inc Supplier No. 7022
Medical grade foam. No adhesives or binders. Non-abradive. High solvent capacity.Little to no particles. Abrasion resistant. ESD Rigid Handle. Wide Paddle Tip. ISO Class 4-7 compatible. Class 10 cleanroom laundered.