Learn More
Electron Microscopy Sciences Polyurethane Foam Swabs
SDP
Catalog No. 501930632
Encompass_Preferred
Durable wrapped foam swabs designed for precise applications provide good particle entrapment and work well as solvent applicators. With the most durable head of any foam swabs, wrapped foam delivers superior abrasion resistance for removal of tough contaminants. Wicking action pulls solvent from surfaces. The small tip allows use in very tight tolerance areas, while the pointed end works especially as pick. Nylon handle.
Catalog No. 50-193-0632 Supplier Electron Microscopy Sciences Supplier No. 7832577
Add to Cart
Add to Cart
Durable wrapped foam swabs designed for precise applications provide good particle entrapment and work well as solvent applicators. With the most durable head of any foam swabs, wrapped foam delivers superior abrasion resistance for removal of tough contaminants. Wicking action pulls solvent from surfaces. The small tip allows use in very tight tolerance areas, while the pointed end works especially as pick. Nylon handle.