Learn More
AVANTOR MASTERFLEX (VWR LLC) Ether-Based Polyurethane Tubing, 1/8 x 1/4"", 100'
Catalog No. NC0421340
Encompass
Ether grade polyurethane has good chemical resistance to weak organic acids and oils, good elongation and recovery. It is nontoxic and transparent. Sterilizable by ethylene oxide.
Catalog No. NC0421340 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9586903
Ether grade polyurethane has good chemical resistance to weak organic acids and oils, good elongation and recovery. It is nontoxic and transparent. Sterilizable by ethylene oxide.