Learn More
Grainger 100 ft. Standard Polyurethane Tubing, 1/4" Outside Dia., 5/32" Inside Dia.
Catalog No. NC1667754
Encompass_Preferred
Strong, flexible tubing is 5 to 10 times more wear- and abrasion-resistant than rubber hose, and lighter weight. Use with push-to-connect fittings with no tube support. Nonmarking; resist algae growth. For air and water lines, robotics, carwash, and painting equipment. Meets NSF 51 standards.
Catalog No. NC1667754 Supplier Grainger Supplier No. 1CTG5
Strong, flexible tubing is 5 to 10 times more wear- and abrasion-resistant than rubber hose, and lighter weight. Use with push-to-connect fittings with no tube support. Nonmarking; resist algae growth. For air and water lines, robotics, carwash, and painting equipment. Meets NSF 51 standards.