Learn More
Enzo Life Sciences POLYVIEW PLUS AP (anti-rabbit) reagent (150 tests)
SDP
Supplier:  Enzo Life Sciences ENZACC1100150
Encompass_Preferred
High sensitivity, low background nanopolymer detection reagent for use with HIGHDEF chromogens in ISH and IHC applications. The POLYVIEW PLUS AP (anti-rabbit) reagent is a ready-to-use, non-biotin, one-step detection reagent suitable for immunohistochemical detection of antigens in formalin-fixed paraffin-embedded tissues and frozen sections. The detection reagent may also be used with blood smears, cytosmears, and cell preparations. In addition, the POLYVIEW PLUS AP (anti-rabbit) reagent can be adapted for in situ hybridization detection. Applications: IHC, ISH (in situ hybridization). Handling: Do not freeze.
Catalog No. NC1609443