Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinyl acetate, M.W. ca 50,000, Thermo Scientific™

Catalog No. p-7020688
Change view
Click to view available options
Quantity:
2500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA127320E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA127320E Supplier Thermo Scientific Chemicals Supplier No. A127320E

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-20-7
Molecular Formula (C4H6O2)n
Molecular Weight (g/mol) 86.09
MDL Number MFCD00084457
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC(-*)C-*

Specifications

Solubility Information Soluble in ketones,ethers and aromatic hydrocarbons.
Quantity 2500 g
Formula Weight ≈50,000
Chemical Name or Material Polyvinyl acetate
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.