Learn More
Electron Microscopy Sciences Polyvinyl Alcohol 15000 500 GR
SDP
Supplier:  Electron Microscopy Sciences 19800
Encompass_Preferred
CAS #9002-89-5 (CH₂CHOH)n F.W. 14000-15000
A water-miscible embedding medium with a degree of Hydrolyzation: 86-89 mol%. See Munoz-Guerra,S. and Subrirana, J.A. (1982). A crosslinked polyvinyl alcohol: a water-miscible polymer used as an embedding medium for electron microscopy. Mikroskopie, 39:346. D
Catalog No. 50-280-30