Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinyl Alcohol 5% Solution, 3.8 L, Innovating Science™
GREENER_CHOICESCI_ED

Catalog No. S001042 Shop All Aldon Products
Change view
Click to view available options
Quantity:
3.8 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001042 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001042 Supplier Aldon Corporation Supplier No. IS25024
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) Mixture

Specifications

Quantity 3.8 L
Percent Purity 5%
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.